Assay Details
| Gene Symbol: LOC4336329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780069.2 | XP_015635555.1 | 4 - 5 | 1017 | 127 | |
| GenBank mRNA | AK241443.1 | - | 4 - 5 | 1012 | 127 | |
More Information
Set Membership: |
|

LOC4336329
Genomic Map