Assay Details
| Gene Symbol: LOC9271676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780021.2 | XP_015635507.1 | 2 - 3 | 707 | 64 | |
| GenBank mRNA | AK072373.1 | - | 2 - 3 | 682 | 64 | |
| AK072585.1 | - | 2 - 3 | 680 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780021.2 transcripts.
Set Membership: |
|

LOC9271676
Genomic Map