Assay Details
Gene Symbol: LOC4336632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778979.2 | XP_015634465.1 | 2 - 3 | 332 | 108 | |
XM_015778980.2 | XP_015634466.1 | 2 - 3 | 332 | 108 | ||
XM_015778981.2 | XP_015634467.1 | 2 - 3 | 332 | 108 | ||
GenBank mRNA | AK064112.1 | - | 2 - 3 | 305 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
