Assay Details
| Gene Symbol: LOC4336726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780024.2 | XP_015635510.1 | 5 - 6 | 846 | 79 | |
| GenBank mRNA | AK110251.1 | - | 3 - 4 | 393 | 79 | |
| AK120348.1 | - | 5 - 6 | 774 | 79 | ||
| BT130504.1 | - | 1 - 2 | 45 | 79 | ||
More Information
Set Membership: |
|

LOC4336726
Genomic Map