Assay Details
| Gene Symbol: LOC4336757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780892.2 | XP_015636378.1 | 1 | 830 | 107 | |
| GenBank mRNA | AK100603.1 | - | 1 | 750 | 107 | |
More Information
Set Membership: |
|

LOC4336757
Genomic Map