Assay Details
Gene Symbol: LOC4336760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015779119.2 | XP_015634605.1 | 2 - 3 | 437 | 100 | |
GenBank mRNA | AK121581.1 | - | 1 - 2 | 56 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
