Assay Details
| Gene Symbol: LOC4336762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778933.2 | XP_015634419.1 | 7 - 8 | 786 | 63 | |
| GenBank mRNA | AK070540.1 | - | 7 - 8 | 685 | 63 | |
| AY551918.1 | - | 7 - 8 | 791 | 63 | ||
| FJ668596.1 | - | 7 - 8 | 719 | 63 | ||
More Information
Set Membership: |
|

LOC4336762
Genomic Map