Assay Details
| Gene Symbol: LOC4336800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779534.2 | XP_015635020.1 | 1 | 1792 | 96 | |
| GenBank mRNA | AK063488.1 | - | 1 | 1347 | 96 | |
More Information
Set Membership: |
|

LOC4336800
Genomic Map