Assay Details
| Gene Symbol: LOC4336921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781794.2 | XP_015637280.1 | 7 - 8 | 800 | 110 | |
| GenBank mRNA | AK060169.1 | - | 7 - 8 | 748 | 110 | |
More Information
Set Membership: |
|

LOC4336921
Genomic Map