Assay Details
Gene Symbol: LOC4336930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781745.2 | 1 - 2 | 507 | 106 | ||
GenBank mRNA | AK065931.1 | - | 1 - 2 | 194 | 106 | |
AK099996.1 | - | 1 - 2 | 115 | 106 | ||
AK121814.1 | - | 1 - 2 | 516 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015781745.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
