Assay Details
Gene Symbol: LOC4336930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781745.2 | 2 - 3 | 656 | 86 | ||
GenBank mRNA | AK065931.1 | - | 2 - 3 | 343 | 86 | |
AK099996.1 | - | 2 - 3 | 264 | 86 | ||
AK121814.1 | - | 2 - 3 | 665 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
