Assay Details
| Gene Symbol: LOC4336930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781745.2 | 6 - 7 | 1204 | 95 | ||
| GenBank mRNA | AK065931.1 | - | 6 - 7 | 891 | 95 | |
| AK099996.1 | - | 6 - 7 | 812 | 95 | ||
| AK121814.1 | - | 6 - 7 | 1213 | 95 | ||
More Information
Set Membership: |
|

LOC4336930
Genomic Map