Assay Details
Gene Symbol: LOC9268788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785188.2 | XP_015640674.1 | 3 - 4 | 830 | 100 | |
GenBank mRNA | AK099734.1 | - | 3 - 4 | 804 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
