Assay Details
| Gene Symbol: LOC4339756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784229.1 | XP_015639715.1 | 1 - 2 | 367 | 66 | |
| GenBank mRNA | AK071721.1 | - | 1 - 2 | 350 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784229.1 transcripts.
Set Membership: |
|

LOC4339756
Genomic Map