Assay Details
Gene Symbol: LOC4337240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780203.2 | XP_015635689.1 | 2 - 3 | 423 | 104 | |
XM_015780204.2 | XP_015635690.1 | 2 - 3 | 421 | 104 | ||
GenBank mRNA | AK059441.1 | - | 2 - 3 | 221 | 104 | |
AK069326.1 | - | 2 - 3 | 224 | 104 | ||
BT129135.1 | - | 2 - 3 | 166 | 104 | ||
BT131105.1 | - | 2 - 3 | 144 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
