Assay Details
Gene Symbol: LOC4337240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780203.2 | XP_015635689.1 | 3 - 4 | 647 | 83 | |
XM_015780204.2 | XP_015635690.1 | 3 - 4 | 645 | 83 | ||
GenBank mRNA | AK059441.1 | - | 3 - 4 | 445 | 83 | |
AK069326.1 | - | 3 - 4 | 448 | 83 | ||
BT129135.1 | - | 3 - 4 | 390 | 83 | ||
BT131105.1 | - | 3 - 4 | 368 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
