Assay Details
| Gene Symbol: LOC4337265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781248.2 | XP_015636734.1 | 2 - 3 | 653 | 76 | |
| XM_015781249.2 | XP_015636735.1 | 2 - 3 | 615 | 76 | ||
| GenBank mRNA | AK068928.1 | - | 2 - 3 | 531 | 76 | |
| AK071255.1 | - | 2 - 3 | 461 | 76 | ||
| HQ324813.1 | - | 2 - 3 | 262 | 76 | ||
More Information
Set Membership: |
|

LOC4337265
Genomic Map