Assay Details
| Gene Symbol: LOC4337265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781248.2 | XP_015636734.1 | 3 - 4 | 704 | 64 | |
| XM_015781249.2 | XP_015636735.1 | 3 - 4 | 666 | 64 | ||
| GenBank mRNA | AK071255.1 | - | 3 - 4 | 512 | 64 | |
| HQ324813.1 | - | 3 - 4 | 313 | 64 | ||
More Information
Set Membership: |
|

LOC4337265
Genomic Map