Assay Details
Gene Symbol: LOC4337265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781248.2 | XP_015636734.1 | 4 - 5 | 873 | 124 | |
XM_015781249.2 | XP_015636735.1 | 4 - 5 | 835 | 124 | ||
GenBank mRNA | AK071255.1 | - | 4 - 5 | 681 | 124 | |
HQ324813.1 | - | 4 - 5 | 482 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
