Assay Details
| Gene Symbol: LOC4337265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781248.2 | XP_015636734.1 | 5 - 6 | 1008 | 135 | |
| XM_015781249.2 | XP_015636735.1 | 5 - 6 | 970 | 135 | ||
| GenBank mRNA | AK071255.1 | - | 5 - 6 | 816 | 135 | |
| HQ324813.1 | - | 5 - 6 | 617 | 135 | ||
More Information
Set Membership: |
|

LOC4337265
Genomic Map