Assay Details
Gene Symbol: LOC4337265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781248.2 | XP_015636734.1 | 7 - 8 | 1206 | 128 | |
XM_015781249.2 | XP_015636735.1 | 7 - 8 | 1168 | 128 | ||
GenBank mRNA | AK068928.1 | - | 3 - 4 | 1669 | 128 | |
AK071255.1 | - | 7 - 8 | 1014 | 128 | ||
HQ324813.1 | - | 7 - 8 | 815 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
