Assay Details
Gene Symbol: LOC4337265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781248.2 | XP_015636734.1 | 8 - 9 | 1470 | 80 | |
GenBank mRNA | AK071255.1 | - | 8 - 9 | 1278 | 80 | |
HQ324813.1 | - | 8 - 9 | 1079 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
