Assay Details
| Gene Symbol: LOC4337389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780505.2 | XP_015635991.1 | 4 - 5 | 2115 | 145 | |
| GenBank mRNA | AK067609.1 | - | 4 - 5 | 2116 | 145 | |
| AK099138.1 | - | 4 - 5 | 2070 | 145 | ||
More Information
Set Membership: |
|

LOC4337389
Genomic Map