Assay Details
Gene Symbol: LOC4337589 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784990.2 | XP_015640476.1 | 1 - 2 | 198 | 92 | |
GenBank mRNA | AK060179.1 | - | 1 - 2 | 204 | 92 | |
AK066175.1 | - | 1 - 2 | 154 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
