Assay Details
Gene Symbol: LOC4337590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784991.2 | XP_015640477.1 | 3 - 4 | 444 | 87 | |
GenBank mRNA | AK121048.1 | - | 3 - 4 | 428 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
