Assay Details
| Gene Symbol: LOC4337722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784055.2 | XP_015639541.1 | 1 - 2 | 185 | 96 | |
| GenBank mRNA | AK058276.1 | - | 1 - 2 | 149 | 96 | |
| AK064884.1 | - | 1 - 2 | 153 | 96 | ||
| AK121623.1 | - | 1 - 2 | 148 | 96 | ||
| DQ060684.1 | - | 1 - 2 | 147 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784055.2 transcripts.
Set Membership: |
|

LOC4337722
Genomic Map