Assay Details
| Gene Symbol: LOC4337722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784055.2 | XP_015639541.1 | 2 - 3 | 375 | 89 | |
| GenBank mRNA | AK058276.1 | - | 2 - 3 | 339 | 89 | |
| AK064884.1 | - | 2 - 3 | 343 | 89 | ||
| DQ060684.1 | - | 2 - 3 | 337 | 89 | ||
More Information
Set Membership: |
|

LOC4337722
Genomic Map