Assay Details
| Gene Symbol: LOC4337722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784055.2 | XP_015639541.1 | 4 - 5 | 524 | 66 | |
| GenBank mRNA | AK058276.1 | - | 4 - 5 | 488 | 66 | |
| AK064884.1 | - | 4 - 5 | 492 | 66 | ||
| AK121623.1 | - | 4 - 5 | 487 | 66 | ||
| DQ060684.1 | - | 4 - 5 | 486 | 66 | ||
More Information
Set Membership: |
|

LOC4337722
Genomic Map