Assay Details
| Gene Symbol: LOC4337722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784055.2 | XP_015639541.1 | 5 - 6 | 804 | 86 | |
| GenBank mRNA | AK058276.1 | - | 5 - 6 | 768 | 86 | |
| AK064884.1 | - | 5 - 6 | 770 | 86 | ||
| AK121623.1 | - | 5 - 6 | 767 | 86 | ||
| DQ060684.1 | - | 5 - 6 | 766 | 86 | ||
More Information
Set Membership: |
|

LOC4337722
Genomic Map