Assay Details
| Gene Symbol: LOC4337722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784055.2 | XP_015639541.1 | 9 - 10 | 1120 | 71 | |
| GenBank mRNA | AK058276.1 | - | 9 - 10 | 1084 | 71 | |
| AK064884.1 | - | 9 - 10 | 1086 | 71 | ||
| AK121623.1 | - | 9 - 10 | 1083 | 71 | ||
More Information
Set Membership: |
|

LOC4337722
Genomic Map