Assay Details
Gene Symbol: LOC4338095 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781955.2 | XP_015637441.1 | 19 - 20 | 3661 | 108 | |
GenBank mRNA | AK110207.1 | - | 1463 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
