Assay Details
Gene Symbol: LOC4338095 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781955.2 | XP_015637441.1 | 1 - 2 | 261 | 65 | |
GenBank mRNA | AK110207.1 | - | 1948 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
