Assay Details
| Gene Symbol: LOC4338095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781955.2 | XP_015637441.1 | 8 - 9 | 1574 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4338095| Gene Symbol: LOC4338095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781955.2 | XP_015637441.1 | 8 - 9 | 1574 | 99 | |
Set Membership: |
|