Assay Details
| Gene Symbol: LOC4338203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782530.2 | XP_015638016.1 | 1 - 2 | 172 | 78 | |
| GenBank mRNA | AK073635.1 | - | 1 - 2 | 94 | 78 | |
| AK104978.1 | - | 1 - 2 | 59 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015782530.2 transcripts.
Set Membership: |
|

LOC4338203
Genomic Map