Assay Details
| Gene Symbol: LOC4338203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782530.2 | XP_015638016.1 | 2 - 3 | 217 | 81 | |
| GenBank mRNA | AK073635.1 | - | 2 - 3 | 139 | 81 | |
| AK104978.1 | - | 2 - 3 | 104 | 81 | ||
More Information
Set Membership: |
|

LOC4338203
Genomic Map