Assay Details
| Gene Symbol: LOC4338348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784704.2 | XP_015640190.1 | 1 - 2 | 181 | 59 | |
| GenBank mRNA | AK067027.1 | - | 1 - 2 | 146 | 59 | |
| AK070329.1 | - | 1 - 2 | 172 | 59 | ||
More Information
Set Membership: |
|

LOC4338348
Genomic Map