Assay Details
| Gene Symbol: LOC4338956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781839.2 | XP_015637325.1 | 4 - 5 | 1056 | 72 | |
| GenBank mRNA | AK106357.1 | - | 4 - 5 | 907 | 72 | |
More Information
Set Membership: |
|

LOC4338956
Genomic Map