Assay Details
| Gene Symbol: LOC4338974 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782934.2 | XP_015638420.1 | 1 - 2 | 548 | 114 | |
| GenBank mRNA | AK101945.1 | - | 1 - 2 | 555 | 114 | |
More Information
Set Membership: |
|

LOC4338974
Genomic Map