Assay Details
Gene Symbol: LOC4339246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781917.2 | XP_015637403.1 | 2 - 3 | 1203 | 72 | |
GenBank mRNA | AK064242.1 | - | 2 - 3 | 1134 | 72 | |
AK065486.1 | - | 2 - 3 | 1119 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
