Assay Details
| Gene Symbol: LOC4339966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785385.2 | XP_015640871.1 | 4 - 5 | 580 | 81 | |
| GenBank mRNA | AK068302.1 | - | 4 - 5 | 586 | 81 | |
| AK104425.1 | - | 4 - 5 | 582 | 81 | ||
| HQ123581.1 | - | 4 - 5 | 582 | 81 | ||
More Information
Set Membership: |
|

LOC4339966
Genomic Map