Assay Details
Gene Symbol: LOC4339966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785385.2 | XP_015640871.1 | 5 - 6 | 631 | 140 | |
GenBank mRNA | AK068302.1 | - | 5 - 6 | 637 | 140 | |
AK104425.1 | - | 5 - 6 | 633 | 140 | ||
HQ123581.1 | - | 5 - 6 | 633 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
