Assay Details
| Gene Symbol: LOC4341453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788133.1 | XP_015643619.1 | 1 - 2 | 186 | 83 | |
| GenBank mRNA | AK071330.1 | - | 1 - 2 | 192 | 83 | |
| AY224485.1 | - | 1 - 2 | 88 | 83 | ||
More Information
Set Membership: |
|

LOC4341453
Genomic Map