Assay Details
| Gene Symbol: LOC4341453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788133.1 | XP_015643619.1 | 3 - 4 | 853 | 65 | |
| GenBank mRNA | AK071330.1 | - | 3 - 4 | 859 | 65 | |
| AY224485.1 | - | 3 - 4 | 755 | 65 | ||
More Information
Set Membership: |
|

LOC4341453
Genomic Map