Assay Details
Gene Symbol: LOC4341531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788511.1 | XP_015643997.1 | 4 - 5 | 1184 | 81 | |
XM_015788512.2 | XP_015643998.1 | 5 - 6 | 1128 | 81 | ||
GenBank mRNA | AK070178.1 | - | 4 - 5 | 1190 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
