Assay Details
Gene Symbol: LOC4341531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788511.1 | XP_015643997.1 | 6 - 7 | 1536 | 128 | |
XM_015788512.2 | XP_015643998.1 | 7 - 8 | 1480 | 128 | ||
GenBank mRNA | AK070178.1 | - | 6 - 7 | 1542 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
