Assay Details
Gene Symbol: LOC4341531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788511.1 | XP_015643997.1 | 8 - 9 | 1735 | 82 | |
XM_015788512.2 | XP_015643998.1 | 9 - 10 | 1679 | 82 | ||
GenBank mRNA | AK070178.1 | - | 8 - 9 | 1741 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
