Assay Details
Gene Symbol: LOC4341607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785701.2 | XP_015641187.1 | 1 | 1262 | 98 | |
GenBank mRNA | AK105817.1 | - | 1 | 1246 | 98 | |
AK121564.1 | - | 1 | 1247 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
