Assay Details
Gene Symbol: LOC4342410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK069726.1 | - | 13 - 14 | 1539 | 69 | |
AK242681.1 | - | 3581 | 69 | |||
BT132595.1 | - | 1 - 2 | 92 | 69 | ||
EU703802.1 | - | 13 - 14 | 1256 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
