Assay Details
| Gene Symbol: LOC4342410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK069726.1 | - | 6 - 7 | 826 | 84 | |
| AK242681.1 | - | 2871 | 84 | |||
| EU703802.1 | - | 6 - 7 | 546 | 84 | ||
More Information
Set Membership: |
|

LOC4342410
Genomic Map