Assay Details
Gene Symbol: LOC4342410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK069726.1 | - | 7 - 8 | 949 | 78 | |
AK242681.1 | - | 2994 | 78 | |||
EU703802.1 | - | 7 - 8 | 669 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
